Learn More
enQuireBio™ Recombinant Mouse Hemoglobin subunit alpha Protein
A DNA sequence encoding the Mus musculus (Mouse) Hemoglobin subunit alpha, was expressed in the hosts and tags indicated.
1820.00 DKK - 24645.00 DKK
Specifications
Name | Hemoglobin subunit alpha Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | Hba |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16061150
|
enQuireBio™
QP6146-ec-10ug |
10 ÎĽg | N/A | ||||||
16001160
|
enQuireBio™
QP6146-ec-50ug |
50 ÎĽg | N/A | ||||||
16051150
|
enQuireBio™
QP6146-ec-100ug |
100 ÎĽg |
5555.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16081150
|
enQuireBio™
QP6146-ec-200ug |
200 ÎĽg | N/A | ||||||
16091150
|
enQuireBio™
QP6146-ec-500ug |
500 ÎĽg | N/A | ||||||
16071150
|
enQuireBio™
QP6146-ec-1mg |
1 mg |
24645.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Hemoglobin subunit alpha Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
Hba | |
Recombinant Protein | |
E. coli | |
VLSGEDKSNI KAAWGKIGGH GAEYGAEALE RMFASFPTTK TYFPHFDVSH GSAQVKGHGK KVADALASAA GHLDDLPGAL SALSDLHAHK LRVDPVNFKL LSHCLLVTLA SHHPADFTPA VHASLDKFLA SVSTVLTSKY R | |
Greater than 80% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
enQuireBio™ Recombinant Mouse Hemoglobin subunit alpha Protein