missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse H2-D1 Protein
A DNA sequence encoding the Mus musculus (Mouse) H2-D1, was expressed in the hosts and tags indicated.
1820.00 DKK - 19665.00 DKK
Specifications
Name | H2-D1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | H2-D1 |
Protein Length | Extracellular Domain |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16098690
|
enQuireBio™
QP7403-ye-10ug |
10 ÎĽg | N/A | ||||||
16038700
|
enQuireBio™
QP7403-ye-50ug |
50 ÎĽg | N/A | ||||||
16088690
|
enQuireBio™
QP7403-ye-100ug |
100 ÎĽg |
5555.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16018700
|
enQuireBio™
QP7403-ye-200ug |
200 ÎĽg | N/A | ||||||
16028700
|
enQuireBio™
QP7403-ye-500ug |
500 ÎĽg |
15875.00 DKK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16008700
|
enQuireBio™
QP7403-ye-1mg |
1 mg | N/A | ||||||
Specifications
H2-D1 Protein | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
H2-D1 | |
Recombinant Protein | |
Yeast | |
GSHSLRYFVT AVSRPGFGEP RYMEVGYVDN TEFVRFDSDA ENPRYEPRAR WIEQEGPEYW ERETRRAKGN EQSFRVDLRT ALRYYNQSAG GSHTLQWMAG CDVESDGRLL RGYWQFAYDG CDYIALNEDL KTWTAADMAA QITRRKWEQA GAAERDRAYL EGECVEWLRR YLKNGNATLL RTDPPKAHVT HHRRPEGDVT LRCWALGFYP ADITLTWQLN GEELTQEMEL VETRPAGDGT FQKWASVVVP LGKEQKYTCH VEHEGLPEPL TLRWGKEEPP SSTKTNT | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse H2-D1 Protein