missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse H2-D1 Protein
A DNA sequence encoding the Mus musculus (Mouse) H2-D1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7403-ye-50ug
This item is not returnable.
View return policy
Specifications
H2-D1 Protein | |
Research Use Only | |
H2-D1 | |
Recombinant Protein | |
Yeast | |
GSHSLRYFVT AVSRPGFGEP RYMEVGYVDN TEFVRFDSDA ENPRYEPRAR WIEQEGPEYW ERETRRAKGN EQSFRVDLRT ALRYYNQSAG GSHTLQWMAG CDVESDGRLL RGYWQFAYDG CDYIALNEDL KTWTAADMAA QITRRKWEQA GAAERDRAYL EGECVEWLRR YLKNGNATLL RTDPPKAHVT HHRRPEGDVT LRCWALGFYP ADITLTWQLN GEELTQEMEL VETRPAGDGT FQKWASVVVP LGKEQKYTCH VEHEGLPEPL TLRWGKEEPP SSTKTNT | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse H2-D1 Protein > 50μg
Spot an opportunity for improvement?Share a Content Correction