missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Galectin-6 Protein

A DNA sequence encoding the Mus musculus (Mouse) Galectin-6, was expressed in the hosts and tags indicated.

1820.00 DKK - 24645.00 DKK

Specifications

Accession Number NP_034837.2
Gene ID (Entrez) 16857
Name Galectin-6 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16052270
View Documents
enQuireBio™
QP6298-ec-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16092270
View Documents
enQuireBio™
QP6298-ec-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16042270
View Documents
enQuireBio™
QP6298-ec-100ug
100 μg
5555.00 DKK
100µg
Please to purchase this item. Need a web account? Register with us today!
16072270
View Documents
enQuireBio™
QP6298-ec-200ug
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16082270
View Documents
enQuireBio™
QP6298-ec-500ug
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16062270
View Documents
enQuireBio™
QP6298-ec-1mg
1 mg
24645.00 DKK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_034837.2
Galectin-6 Protein
Not determined.
Full Length
Mouse
GST
Tris-based buffer, 50% glycerol
16857
Research Use Only
Lgals6
Recombinant Protein
E. coli
MAYVPAPGYQ PTYNPTLPYK RPIPGGLSVG MSFYIQGTAK ENMRRFHVNF AVGQDDGADV AFHFNPRFDG WDKVVFNTKQ SGRWGKEEEK SMPFQKGKHF ELVFMVMPEH YKVVVNGSPF YEYGHRLPVQ MVTHLQVDGD LELQSINFFG VQPAETKYPA MTGPPVFNPC LPYVGALQGG FTVRRTIIIK GYVLPTAKTF AINFRVGSSE DIALHINPRI GDCLVRNSYM NGSWGTEERM VAYNPFGPGQ FFDLSIRCGM DRFKVFANGI HLFNFSHRFQ ALRKINTLEI NGDLTLSYVH I
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse Galectin-6 Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.