missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TNF receptor-associated factor 6 Protein

A DNA sequence encoding the Homo sapiens (Human) TNF receptor-associated factor 6, was expressed in the hosts and tags indicated.

1185.00 DKK - 17250.00 DKK

Specifications

Accession Number NP_004611.1
Gene ID (Entrez) 7189
Name TNF receptor-associated factor 6 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16086190
View Documents
enQuireBio™
QP6838-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16026200
View Documents
enQuireBio™
QP6838-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16076190
View Documents
enQuireBio™
QP6838-ec-100ug
100 ÎĽg
3825.00 DKK
100µg
Please to purchase this item. Need a web account? Register with us today!
16006200
View Documents
enQuireBio™
QP6838-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16016200
View Documents
enQuireBio™
QP6838-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16096190
View Documents
enQuireBio™
QP6838-ec-1mg
1 mg
17250.00 DKK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_004611.1
TNF receptor-associated factor 6 Protein
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
7189
Research Use Only
TRAF6
Recombinant Protein
E. coli
MSLLNCENSC GSSQSESDCC VAMASSCSAV TKDDSVGGTA STGNLSSSFM EEIQGYDVEF DPPLESKYEC PICLMALREA VQTPCGHRFC KACIIKSIRD AGHKCPVDNE ILLENQLFPD NFAKREILSL MVKCPNEGCL HKMELRHLED HQAHCEFALM DCPQCQRPFQ KFHINIHILK DCPRRQVSCD NCAASMAFED KEIHDQNCPL ANVICEYCNT ILIREQMPNH YDLDCPTAPI PCTFSTFGCH EKMQRNHLAR HLQENTQSHM RMLAQAVHSL SVIPDSGYIS EVRNFQETIH QLEGRLVRQD HQIRELTAKM ETQSMYVSEL KRTIRTLEDK VAEIEAQQCN GIYIWKIGNF GMHLKCQEEE KPVVIHSPGF YTGKPGYKLC MRLHLQLPTA QRCANYISLF VHTMQGEYDS HLPWPFQGTI RLTILDQSEA PVRQNHEEIM DAKPELLAFQ RPTIPRNPKG FGYVTFMHLE ALRQRTFIKD DTLLVRCEVS TRFDMGSLRR EGFQPRSTDA GV
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human TNF receptor-associated factor 6 Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.