missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TLR2 / CD282 Protein
A DNA sequence encoding the Homo sapiens (Human) TLR2 / CD282, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6800-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001305716.1 | |
TLR2 / CD282 Protein | |
Research Use Only | |
TLR2 | |
Recombinant Protein | |
E. coli | |
KEESSNQASL SCDRNGICKG SSGSLNSIPS GLTEAVKSLD LSNNRITYIS NSDLQRCVNL QALVLTSNGI NTIEEDSFSS LGSLEHLDLS YNYLSNLSSS WFKPLSSLTF LNLLGNPYKT LGETSLFSHL TKLQILRVGN MDTFTKIQRK DFAGLTFLEE LEIDASDLQS YEPKSLKSIQ NVSHLILHMK QHILLLEIFV DVTSSVECLE LRDTDLDTFH FSELSTGETN SLIKKFTFRN VKITDESLFQ VMKLLNQISG LLELEFDDCT LNGVGNFRAS DNDRVIDPGK VETLTIRRLH IPRFYLFYDL STLYSLTERV KRITVENSKV FLVPCLLSQH LKSLEYLDLS ENLMVEEYLK NSACEDAWPS LQTLILRQNH LASLEKTGET LLTLKNLTNI DISKNSFHSM PETCQWPEKM KYLNLSSTRI HSVTGCIPKT LEILDVSNNN LNLFSLNLPQ LKELYISRNK LMTLPDASLL PMLLVLKISR NAITTFSKEQ LDSFHTLKTL EAGGNNFICS CEFLSFTQEQ QALAKVLIDW PANYLCDSPS HVRGQQVQDV RLSVSECHRT | |
Greater than 90% as determined by SDS-PAGE. |
7097 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |