Learn More
enQuireBio™ Recombinant Human Serine protease HTRA1 Protein
A DNA sequence encoding the Homo sapiens (Human) Serine protease HTRA1, was expressed in the hosts and tags indicated.
1185.00 DKK - 15565.00 DKK
Specifications
Accession Number | NP_002766.1 |
---|---|
Gene ID (Entrez) | 5654 |
Name | Serine protease HTRA1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16061251
|
enQuireBio™
QP7864-EC-100UG |
100 μg |
3450.00 DKK
100µg |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16081251
|
enQuireBio™
QP7864-EC-1MG |
1 mg |
15565.00 DKK
1mg |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16071251
|
enQuireBio™
QP7864-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16091251
|
enQuireBio™
QP7864-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001261
|
enQuireBio™
QP7864-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16011261
|
enQuireBio™
QP7864-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002766.1 | |
Serine protease HTRA1 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
5654 | |
Research Use Only | |
HTRA1 | |
Recombinant Protein | |
E. coli | |
QLSRAGRSAP LAAGCPDRCE PARCPPQPEH CEGGRARDAC GCCEVCGAPE GAACGLQEGP CGEGLQCVVP FGVPASATVR RRAQAGLCVC ASSEPVCGSD ANTYANLCQL RAASRRSERL HRPPVIVLQR GACGQGQEDP NSLRHKYNFI ADVVEKIAPA VVHIELFRKL PFSKREVPVA SGSGFIVSED GLIVTNAHVV TNKHRVKVEL KNGATYEAKI KDVDEKADIA LIKIDHQGKL PVLLLGRSSE LRPGEFVVAI GSPFSLQNTV TTGIVSTTQR GGKELGLRNS DMDYIQTDAI INYGNSGGPL VNLDGEVIGI NTLKVTAGIS FAIPSDKIKK FLTESHDRQA KGKAITKKKY IGIRMMSLTS SKAKELKDRH RDFPDVISGA YIIEVIPDTP AEAGGLKEND VIISINGQSV VSANDVSDVI KRESTLNMVV RRGNEDIMIT VIPEEIDP | |
Greater than 90% as determined by SDS-PAGE. |