missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Serine / arginine-rich splicing factor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Serine/arginine-rich splicing factor 1, was expressed in the hosts and tags indicated.
1185.00 DKK - 17250.00 DKK
Specifications
Accession Number | NP_001071634.1 |
---|---|
Gene ID (Entrez) | 6426 |
Name | Serine / arginine-rich splicing factor 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16075030
|
enQuireBio™
QP6679-ec-10ug |
10 ÎĽg | N/A | ||||||
16015040
|
enQuireBio™
QP6679-ec-50ug |
50 ÎĽg | N/A | ||||||
16065030
|
enQuireBio™
QP6679-ec-100ug |
100 ÎĽg |
3825.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16095030
|
enQuireBio™
QP6679-ec-200ug |
200 ÎĽg | N/A | ||||||
16005040
|
enQuireBio™
QP6679-ec-500ug |
500 ÎĽg | N/A | ||||||
16085030
|
enQuireBio™
QP6679-ec-1mg |
1 mg |
17250.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001071634.1 | |
Serine / arginine-rich splicing factor 1 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
6426 | |
Research Use Only | |
SRSF1 | |
Recombinant Protein | |
E. coli | |
SGGGVIRGPA GNNDCRIYVG NLPPDIRTKD IEDVFYKYGA IRDIDLKNRR GGPPFAFVEF EDPRDAEDAV YGRDGYDYDG YRLRVEFPRS GRGTGRGGGG GGGGGAPRGR YGPPSRRSEN RVVVSGLPPS GSWQDLKDHM REAGDVCYAD VYRDGTGVVE FVRKEDMTYA VRKLDNTKFR SHEGETAYIR VKVDGPRSPS YGRSRSRSRS RSRSRSRSNS RSRSYSPRRS RGSPRYSPRH SRSRSRT | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Serine / arginine-rich splicing factor 1 Protein