missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Serine / arginine-rich splicing factor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Serine/arginine-rich splicing factor 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6679-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_001071634.1 | |
Serine / arginine-rich splicing factor 1 Protein | |
Research Use Only | |
SRSF1 | |
Recombinant Protein | |
E. coli | |
SGGGVIRGPA GNNDCRIYVG NLPPDIRTKD IEDVFYKYGA IRDIDLKNRR GGPPFAFVEF EDPRDAEDAV YGRDGYDYDG YRLRVEFPRS GRGTGRGGGG GGGGGAPRGR YGPPSRRSEN RVVVSGLPPS GSWQDLKDHM REAGDVCYAD VYRDGTGVVE FVRKEDMTYA VRKLDNTKFR SHEGETAYIR VKVDGPRSPS YGRSRSRSRS RSRSRSRSNS RSRSYSPRRS RGSPRYSPRH SRSRSRT | |
Greater than 90% as determined by SDS-PAGE. |
6426 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Serine / arginine-rich splicing factor 1 Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction