missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ribose-phosphate pyrophosphokinase 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Ribose-phosphate pyrophosphokinase 1, was expressed in the hosts and tags indicated.

1185.00 DKK - 17250.00 DKK

Specifications

Accession Number NP_001191331.1
Gene ID (Entrez) 5631
Name Ribose-phosphate pyrophosphokinase 1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16084000
View Documents
enQuireBio™
QP6535-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16024010
View Documents
enQuireBio™
QP6535-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16074000
View Documents
enQuireBio™
QP6535-ec-100ug
100 ÎĽg
3825.00 DKK
100µg
Please to purchase this item. Need a web account? Register with us today!
16004010
View Documents
enQuireBio™
QP6535-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16014010
View Documents
enQuireBio™
QP6535-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16094000
View Documents
enQuireBio™
QP6535-ec-1mg
1 mg
17250.00 DKK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_001191331.1
Ribose-phosphate pyrophosphokinase 1 Protein
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
5631
Research Use Only
PRPS1
Recombinant Protein
E. coli
PNIKIFSGSS HQDLSQKIAD RLGLELGKVV TKKFSNQETC VEIGESVRGE DVYIVQSGCG EINDNLMELL IMINACKIAS ASRVTAVIPC FPYARQDKKD KSRAPISAKL VANMLSVAGA DHIITMDLHA SQIQGFFDIP VDNLYAEPAV LKWIRENISE WRNCTIVSPD AGGAKRVTSI ADRLNVDFAL IHKERKKANE VDRMVLVGDV KDRVAILVDD MADTCGTICH AADKLLSAGA TRVYAILTHG IFSGPAISRI NNACFEAVVV TNTIPQEDKM KHCSKIQVID ISMILAEAIR RTHNGESVSY LFSHVPL
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human Ribose-phosphate pyrophosphokinase 1 Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.