missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ribose-phosphate pyrophosphokinase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Ribose-phosphate pyrophosphokinase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6535-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_001191331.1 | |
Ribose-phosphate pyrophosphokinase 1 Protein | |
Research Use Only | |
PRPS1 | |
Recombinant Protein | |
E. coli | |
PNIKIFSGSS HQDLSQKIAD RLGLELGKVV TKKFSNQETC VEIGESVRGE DVYIVQSGCG EINDNLMELL IMINACKIAS ASRVTAVIPC FPYARQDKKD KSRAPISAKL VANMLSVAGA DHIITMDLHA SQIQGFFDIP VDNLYAEPAV LKWIRENISE WRNCTIVSPD AGGAKRVTSI ADRLNVDFAL IHKERKKANE VDRMVLVGDV KDRVAILVDD MADTCGTICH AADKLLSAGA TRVYAILTHG IFSGPAISRI NNACFEAVVV TNTIPQEDKM KHCSKIQVID ISMILAEAIR RTHNGESVSY LFSHVPL | |
Greater than 90% as determined by SDS-PAGE. |
5631 | |
50 ÎĽg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Ribose-phosphate pyrophosphokinase 1 Protein > 50μg
Spot an opportunity for improvement?Share a Content Correction