missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ras association domain-containing protein 1

A DNA sequence encoding the Homo sapiens (Human) Ras association domain-containing protein 6, was expressed in the hosts and tags indicated.

1500.00 DKK - 20535.00 DKK

Specifications

Accession Number NP_001257320.1
Gene ID (Entrez) 166824
Name Ras association domain-containing protein 1
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16000661
View Documents
enQuireBio™
QP7763-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16040661
View Documents
enQuireBio™
QP7763-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16090651
View Documents
enQuireBio™
QP7763-ec-100ug
100 ÎĽg
4735.00 DKK
100µg
Please to purchase this item. Need a web account? Register with us today!
16020661
View Documents
enQuireBio™
QP7763-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16030661
View Documents
enQuireBio™
QP7763-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16010661
View Documents
enQuireBio™
QP7763-ec-1mg
1 mg
20535.00 DKK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_001257320.1
Ras association domain-containing protein 1
Not determined.
Full Length of Isoform 4
Human
GST
Tris-based buffer, 50% glycerol
166824
Research Use Only
RASSF6
Recombinant Protein
E. coli
MTMMAHQYPS WIFINEKTFI TREQLNSLLK TYNIFYENQK NLHILYGETE DGKLIVEGML DIFWGVKRPI QLKIQDEKPF SSFTSMKSSD VFSSKGMTRW GEFDDLYRIS ELDRTQIPMS EKRNSQEDYL SYHSNTLKPH AKDEPDSPVL YRTMSEAALV RKRMKPLMMD RKERQKNRAS INGHFYNHEI ENSPQDFALH IIFATGEQRR LKKTDIPLLQ RLLQGPSEKN ARIFLMDKDA EEISSDVAQY INFHFSLLES ILQRLNEEEK REIQRIVTKF NKEKAIILKC LQNKLVIKTE TTV
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human Ras association domain-containing protein 1

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.