missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ras association domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) Ras association domain-containing protein 6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7763-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001257320.1 | |
Ras association domain-containing protein 1 | |
Research Use Only | |
RASSF6 | |
Recombinant Protein | |
E. coli | |
MTMMAHQYPS WIFINEKTFI TREQLNSLLK TYNIFYENQK NLHILYGETE DGKLIVEGML DIFWGVKRPI QLKIQDEKPF SSFTSMKSSD VFSSKGMTRW GEFDDLYRIS ELDRTQIPMS EKRNSQEDYL SYHSNTLKPH AKDEPDSPVL YRTMSEAALV RKRMKPLMMD RKERQKNRAS INGHFYNHEI ENSPQDFALH IIFATGEQRR LKKTDIPLLQ RLLQGPSEKN ARIFLMDKDA EEISSDVAQY INFHFSLLES ILQRLNEEEK REIQRIVTKF NKEKAIILKC LQNKLVIKTE TTV | |
Greater than 80% as determined by SDS-PAGE. |
166824 | |
1 mg | |
Not determined. | |
Full Length of Isoform 4 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |