missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein quaking Protein
A DNA sequence encoding the Homo sapiens (Human) Protein quaking, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7918-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001288014.1 | |
Protein quaking Protein | |
Research Use Only | |
QKI | |
Recombinant Protein | |
E. coli | |
MVGEMETKEK PKPTPDYLMQ LMNDKKLMSS LPNFCGIFNH LERLLDEEIS RVRKDMYNDT LNGSTEKRSA ELPDAVGPIV QLQEKLYVPV KEYPDFNFVG RILGPRGLTA KQLEAETGCK IMVRGKGSMR DKKKEEQNRG KPNWEHLNED LHVLITVEDA QNRAEIKLKR AVEEVKKLLV PAAEGEDSLK KMQLMELAIL NGTYRDANIK SPALAFSLAA TAQAAPRIIT GPAPVLPPAA LRTPTPAGPT IMPLIRQIQT AVMPNGTPHP TAAIVPPGPE AGLIYTPYEY PYTLAPATSI LEYPIEPSGV LGAVATKVRR HDMRVHPYQR IVTADRAATG N | |
Greater than 80% as determined by SDS-PAGE. |
9444 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |