missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRKAG2 Protein
A DNA sequence encoding the Homo sapiens (Human) PRKAG2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8207-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001035723.1 | |
PRKAG2 Protein | |
Research Use Only | |
PRKAG2 | |
Recombinant Protein | |
E. coli | |
MLEKLEFEDE AVEDSESGVY MRFMRSHKCY DIVPTSSKLV VFDTTLQVKK AFFALVANGV RAAPLWESKK QSFVGMLTIT DFINILHRYY KSPMVQIYEL EEHKIETWRE LYLQETFKPL VNISPDASLF DAVYSLIKNK IHRLPVIDPI SGNALYILTH KRILKFLQLF MSDMPKPAFM KQNLDELGIG TYHNIAFIHP DTPIIKALNI FVERRISALP VVDESGKVVD IYSKFDVINL AAEKTYNNLD ITVTQALQHR SQYFEGVVKC NKLEILETIV DRIVRAEVHR LVVVNEADSI VGIISLSDIL QALILTPAGA KQKETETE | |
Greater than 90% as determined by SDS-PAGE. |
51422 | |
10 μg | |
Not determined. | |
Full Length of Isoform B | |
Human | |
His | |
Tris-based buffer, 50% glycerol |