missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PI3 / Elafin / WFDC14 Protein
A DNA sequence encoding the Homo sapiens (Human) PI3 / Elafin / WFDC14, was expressed in the hosts and tags indicated.
1185.00 DKK - 15565.00 DKK
Specifications
Accession Number | NP_002629.1 |
---|---|
Gene ID (Entrez) | 5266 |
Name | PI3 / Elafin / WFDC14 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16083690
|
enQuireBio™
QP6493-EC-100UG |
100 μg |
3450.00 DKK
100µg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16003700
|
enQuireBio™
QP6493-EC-1MG |
1 mg |
15565.00 DKK
1mg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16093690
|
enQuireBio™
QP6493-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16013700
|
enQuireBio™
QP6493-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16023700
|
enQuireBio™
QP6493-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033700
|
enQuireBio™
QP6493-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002629.1 | |
PI3 / Elafin / WFDC14 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
5266 | |
Research Use Only | |
PI3 | |
Recombinant Protein | |
E. coli | |
AQEPVKGPVS TKPGSCPIIL IRCAMLNPPN RCLKDTDCPG IKKCCEGSCG MACFVPQ | |
Greater than 90% as determined by SDS-PAGE. |