missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Odontogenic ameloblast-associated Protein
A DNA sequence encoding the Homo sapiens (Human) Odontogenic ameloblast-associated protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6445-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_060325.3 | |
Odontogenic ameloblast-associated Protein | |
Research Use Only | |
ODAM | |
Recombinant Protein | |
E. coli | |
APLIPQRLMS ASNSNELLLN LNNGQLLPLQ LQGPLNSWIP PFSGILQQQQ QAQIPGLSQF SLSALDQFAG LLPNQIPLTG EASFAQGAQA GQVDPLQLQT PPQTQPGPSH VMPYVFSFKM PQEQGQMFQY YPVYMVLPWE QPQQTVPRSP QQTRQQQYEE QIPFYAQFGY IPQLAEPAIS GGQQQLAFDP QLGTAPEIAV MSTGEEIPYL QKEAINFRHD SAGVFMPSTS PKPSTTNVFT SAVDQTITPE LPEEKDKTDS LREP | |
Greater than 80% as determined by SDS-PAGE. |
54959 | |
1 mg | |
Not determined. | |
Full Length of Mature. Protein | |
Human | |
His | |
Tris-based buffer, 50% glycerol |