missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Odontogenic ameloblast-associated Protein

A DNA sequence encoding the Homo sapiens (Human) Odontogenic ameloblast-associated protein, was expressed in the hosts and tags indicated.

1185.00 DKK - 15565.00 DKK

Specifications

Accession Number NP_060325.3
Gene ID (Entrez) 54959
Name Odontogenic ameloblast-associated Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16063360
View Documents Promotion Details
enQuireBio™
QP6445-EC-100UG
100 μg
3450.00 DKK
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 18-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16083360
View Documents Promotion Details
enQuireBio™
QP6445-EC-1MG
1 mg
15565.00 DKK
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 18-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16073360
View Documents Promotion Details
enQuireBio™
QP6445-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16093360
View Documents Promotion Details
enQuireBio™
QP6445-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16003370
View Documents Promotion Details
enQuireBio™
QP6445-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16013370
View Documents Promotion Details
enQuireBio™
QP6445-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_060325.3
Odontogenic ameloblast-associated Protein
Not determined.
Full Length of Mature. Protein
Human
His
Tris-based buffer, 50% glycerol
54959
Research Use Only
ODAM
Recombinant Protein
E. coli
APLIPQRLMS ASNSNELLLN LNNGQLLPLQ LQGPLNSWIP PFSGILQQQQ QAQIPGLSQF SLSALDQFAG LLPNQIPLTG EASFAQGAQA GQVDPLQLQT PPQTQPGPSH VMPYVFSFKM PQEQGQMFQY YPVYMVLPWE QPQQTVPRSP QQTRQQQYEE QIPFYAQFGY IPQLAEPAIS GGQQQLAFDP QLGTAPEIAV MSTGEEIPYL QKEAINFRHD SAGVFMPSTS PKPSTTNVFT SAVDQTITPE LPEEKDKTDS LREP
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications