missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human N-myc proto-oncogene Protein

A DNA sequence encoding the Homo sapiens (Human) N-myc proto-oncogene protein, was expressed in the hosts and tags indicated.

1500.00 DKK - 15995.00 DKK

Specifications

Accession Number NP_001280157.1
Gene ID (Entrez) 4613
Name N-myc proto-oncogene Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16088031
View Documents
enQuireBio™
QP9298-ye-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16028041
View Documents
enQuireBio™
QP9298-ye-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16078031
View Documents
enQuireBio™
QP9298-ye-100ug
100 ÎĽg
4645.00 DKK
100µg
Please to purchase this item. Need a web account? Register with us today!
16008041
View Documents
enQuireBio™
QP9298-ye-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16018041
View Documents
enQuireBio™
QP9298-ye-500ug
500 ÎĽg
12590.00 DKK
500µg
Please to purchase this item. Need a web account? Register with us today!
16098031
View Documents
enQuireBio™
QP9298-ye-1mg
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

NP_001280157.1
N-myc proto-oncogene Protein
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
4613
Research Use Only
MYCN
Recombinant Protein
Yeast
MPSCSTSTMP GMICKNPDLE FDSLQPCFYP DEDDFYFGGP DSTPPGEDIW KKFELLPTPP LSPSRGFAEH SSEPPSWVTE MLLENELWGS PAEEDAFGLG GLGGLTPNPV ILQDCMWSGF SAREKLERAV SEKLQHGRGP PTAGSTAQSP GAGAASPAGR GHGGAAGAGR AGAALPAELA HPAAECVDPA VVFPFPVNKR EPAPVPAAPA SAPAAGPAVA SGAGIAAPAG APGVAPPRPG GRQTSGGDHK ALSTSGEDTL SDSDDEDDEE EDEEEEIDVV TVEKRRSSSN TKAVTTFTIT VRPKNAALGP GRAQSSELIL KRCLPIHQQH NYAAPSPYVE SEDAPPQKKI KSEASPRPLK SVIPPKAKSL SPRNSDSEDS ERRRNHNILE RQRRNDLRSS FLTLRDHVPE LVKNEKAAKV VILKKATEYV HSLQAEEHQL LLEKEKLQAR QQQLLKKIEH ARTC
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human N-myc proto-oncogene Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.