Learn More
enQuireBio™ Recombinant Human N-myc proto-oncogene Protein
A DNA sequence encoding the Homo sapiens (Human) N-myc proto-oncogene protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9298-ye-10ug
This item is not returnable.
View return policy
Specifications
NP_001280157.1 | |
N-myc proto-oncogene Protein | |
Research Use Only | |
MYCN | |
Recombinant Protein | |
Yeast | |
MPSCSTSTMP GMICKNPDLE FDSLQPCFYP DEDDFYFGGP DSTPPGEDIW KKFELLPTPP LSPSRGFAEH SSEPPSWVTE MLLENELWGS PAEEDAFGLG GLGGLTPNPV ILQDCMWSGF SAREKLERAV SEKLQHGRGP PTAGSTAQSP GAGAASPAGR GHGGAAGAGR AGAALPAELA HPAAECVDPA VVFPFPVNKR EPAPVPAAPA SAPAAGPAVA SGAGIAAPAG APGVAPPRPG GRQTSGGDHK ALSTSGEDTL SDSDDEDDEE EDEEEEIDVV TVEKRRSSSN TKAVTTFTIT VRPKNAALGP GRAQSSELIL KRCLPIHQQH NYAAPSPYVE SEDAPPQKKI KSEASPRPLK SVIPPKAKSL SPRNSDSEDS ERRRNHNILE RQRRNDLRSS FLTLRDHVPE LVKNEKAAKV VILKKATEYV HSLQAEEHQL LLEKEKLQAR QQQLLKKIEH ARTC | |
Greater than 90% as determined by SDS-PAGE. |
4613 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Product Title
enQuireBio™ Recombinant Human N-myc proto-oncogene Protein > 10μg
Spot an opportunity for improvement?Share a Content Correction