missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human JmjC domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) JmjC domain-containing protein 7, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6248-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001108104.1 | |
JmjC domain-containing protein 1 | |
Research Use Only | |
JMJD7 | |
Recombinant Protein | |
E. coli | |
MAEAALEAVR SELREFPAAA RELCVPLAVP YLDKPPTPLH FYRDWVCPNR PCIIRNALQH WPALQKWSLP YFRATVGSTE VSVAVTPDGY ADAVRGDRFM MPAERRLPLS FVLDVLEGRA QHPGVLYVQK QCSNLPSELP QLLPDLESHV PWASEALGKM PDAVNFWLGE AAAVTSLHKD HYENLYCVVS GEKHFLFHPP SDRPFIPYEL YTPATYQLTE EGTFKVVDEE AMEKVPWIPL DPLAPDLARY PSYSQAQALR CTVRAGEMLY LPALWFHHVQ QSQGCIAVNF WYDMEYDLKY SYFQLLDSLT KASGLD | |
Greater than 90% as determined by SDS-PAGE. |
100137047 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |