missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human JmjC domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) JmjC domain-containing protein 7, was expressed in the hosts and tags indicated.
1185.00 DKK - 17250.00 DKK
Specifications
Accession Number | NP_001108104.1 |
---|---|
Gene ID (Entrez) | 100137047 |
Name | JmjC domain-containing protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16081910
|
enQuireBio™
QP6248-ec-10ug |
10 ÎĽg | N/A | ||||||
16021920
|
enQuireBio™
QP6248-ec-50ug |
50 ÎĽg | N/A | ||||||
16071910
|
enQuireBio™
QP6248-ec-100ug |
100 ÎĽg |
3825.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16001920
|
enQuireBio™
QP6248-ec-200ug |
200 ÎĽg | N/A | ||||||
16011920
|
enQuireBio™
QP6248-ec-500ug |
500 ÎĽg | N/A | ||||||
16091910
|
enQuireBio™
QP6248-ec-1mg |
1 mg |
17250.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001108104.1 | |
JmjC domain-containing protein 1 | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
100137047 | |
Research Use Only | |
JMJD7 | |
Recombinant Protein | |
E. coli | |
MAEAALEAVR SELREFPAAA RELCVPLAVP YLDKPPTPLH FYRDWVCPNR PCIIRNALQH WPALQKWSLP YFRATVGSTE VSVAVTPDGY ADAVRGDRFM MPAERRLPLS FVLDVLEGRA QHPGVLYVQK QCSNLPSELP QLLPDLESHV PWASEALGKM PDAVNFWLGE AAAVTSLHKD HYENLYCVVS GEKHFLFHPP SDRPFIPYEL YTPATYQLTE EGTFKVVDEE AMEKVPWIPL DPLAPDLARY PSYSQAQALR CTVRAGEMLY LPALWFHHVQ QSQGCIAVNF WYDMEYDLKY SYFQLLDSLT KASGLD | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human JmjC domain-containing protein 1