Learn More
enQuireBio™ Recombinant Human Integrin alpha-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Integrin alpha-2, was expressed in the hosts and tags indicated.
1500.00 DKK - 15995.00 DKK
Specifications
Accession Number | NP_002194.2 |
---|---|
Gene ID (Entrez) | 3673 |
Name | Integrin alpha-2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16031830
|
enQuireBio™
QP6238-YE-100UG |
100 μg |
4190.00 DKK
100µg |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16071830
|
enQuireBio™
QP6238-YE-500UG |
500 μg |
11360.00 DKK
500µg |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16041830
|
enQuireBio™
QP6238-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061830
|
enQuireBio™
QP6238-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081830
|
enQuireBio™
QP6238-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16051830
|
enQuireBio™
QP6238-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002194.2 | |
Integrin alpha-2 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3673 | |
Research Use Only | |
ITGA2 | |
Recombinant Protein | |
Yeast | |
WDAVKNFLEK FVQGLDIGPT KTQVGLIQYA NNPRVVFNLN TYKTKEEMIV ATSQTSQYGG DLTNTFGAIQ YARKYAYSAA SGGRRSATKV MVVVTDGESH DGSMLKAVID QCNHDNILRF GIAVLGYLNR NALDTKNLIK EIKAIASIPT ERYFFNVSDE AALLEKAGTL GEQIFSIE | |
Greater than 90% as determined by SDS-PAGE. |