missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ig gamma-1 chain C region Protein
A DNA sequence encoding the Homo sapiens (Human) Ig gamma-1 chain C region, was expressed in the hosts and tags indicated.
1185.00 DKK - 17250.00 DKK
Specifications
Name | Ig gamma-1 chain C region Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | IGHG1 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16081580
|
enQuireBio™
QP6209-ec-10ug |
10 ÎĽg | N/A | ||||||
16021590
|
enQuireBio™
QP6209-ec-50ug |
50 ÎĽg | N/A | ||||||
16071580
|
enQuireBio™
QP6209-ec-100ug |
100 ÎĽg |
3825.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16001590
|
enQuireBio™
QP6209-ec-200ug |
200 ÎĽg | N/A | ||||||
16011590
|
enQuireBio™
QP6209-ec-500ug |
500 ÎĽg | N/A | ||||||
16091580
|
enQuireBio™
QP6209-ec-1mg |
1 mg |
17250.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Ig gamma-1 chain C region Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
IGHG1 | |
Recombinant Protein | |
E. coli | |
ASTKGPSVFP LAPSSKSTSG GTAALGCLVK DYFPEPVTVS WNSGALTSGV HTFPAVLQSS GLYSLSSVVT VPSSSLGTQT YICNVNHKPS NTKVDKKVEP KSCDKTHTCP PCPAPELLGG PSVFLFPPKP KDTLMISRTP EVTCVVVDVS HEDPEVKFNW YVDGVEVHNA KTKPREEQYN STYRVVSVLT VLHQDWLNGK EYKCKVSNKA LPAPIEKTIS KAKGQPREPQ VYTLPPSRDE LTKNQVSLTC LVKGFYPSDI AVEWESNGQP ENNYKTTPPV LDSDGSFFLY SKLTVDKSRW QQGNVFSCSV MHEALHNHYT QKSLSLSPGK | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Ig gamma-1 chain C region Protein