missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ig gamma-1 chain C region Protein
A DNA sequence encoding the Homo sapiens (Human) Ig gamma-1 chain C region, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6209-ec-10ug
This item is not returnable.
View return policy
Specifications
Ig gamma-1 chain C region Protein | |
Research Use Only | |
IGHG1 | |
Recombinant Protein | |
E. coli | |
ASTKGPSVFP LAPSSKSTSG GTAALGCLVK DYFPEPVTVS WNSGALTSGV HTFPAVLQSS GLYSLSSVVT VPSSSLGTQT YICNVNHKPS NTKVDKKVEP KSCDKTHTCP PCPAPELLGG PSVFLFPPKP KDTLMISRTP EVTCVVVDVS HEDPEVKFNW YVDGVEVHNA KTKPREEQYN STYRVVSVLT VLHQDWLNGK EYKCKVSNKA LPAPIEKTIS KAKGQPREPQ VYTLPPSRDE LTKNQVSLTC LVKGFYPSDI AVEWESNGQP ENNYKTTPPV LDSDGSFFLY SKLTVDKSRW QQGNVFSCSV MHEALHNHYT QKSLSLSPGK | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Ig gamma-1 chain C region Protein > 10μg
Spot an opportunity for improvement?Share a Content Correction