missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Golgi SNAP receptor complex member 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Golgi SNAP receptor complex member 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6107-ec-100ug
This item is not returnable.
View return policy
Specifications
NP_001012529.1 | |
Golgi SNAP receptor complex member 2 Protein | |
Research Use Only | |
GOSR2 | |
Recombinant Protein | |
E. coli | |
MDPLFQQTHK QVHEIQSCMG RLETADKQSV HIVENEIQAS IDQIFSRLER LEILSSKEPP NKRQNARLRV DQLKYDVQHL QTALRNFQHR RHAREQQERQ REELLSRTFT TNDSDTTIPM DESLQFNSSL QKVHNGMDDL ILDGHNILDG LRTQRLTLKG TQKKILDIAN MLGLSNTVMR LIEKRAFQDK | |
Greater than 90% as determined by SDS-PAGE. |
9570 | |
100 ÎĽg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Golgi SNAP receptor complex member 2 Protein > 100μg
Spot an opportunity for improvement?Share a Content Correction