missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Golgi SNAP receptor complex member 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Golgi SNAP receptor complex member 2, was expressed in the hosts and tags indicated.
1185.00 DKK - 17250.00 DKK
Specifications
Accession Number | NP_001012529.1 |
---|---|
Gene ID (Entrez) | 9570 |
Name | Golgi SNAP receptor complex member 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16000850
|
enQuireBio™
QP6107-ec-10ug |
10 ÎĽg | N/A | ||||||
16040850
|
enQuireBio™
QP6107-ec-50ug |
50 ÎĽg | N/A | ||||||
16090840
|
enQuireBio™
QP6107-ec-100ug |
100 ÎĽg |
3825.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16020850
|
enQuireBio™
QP6107-ec-200ug |
200 ÎĽg | N/A | ||||||
16030850
|
enQuireBio™
QP6107-ec-500ug |
500 ÎĽg | N/A | ||||||
16010850
|
enQuireBio™
QP6107-ec-1mg |
1 mg |
17250.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001012529.1 | |
Golgi SNAP receptor complex member 2 Protein | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
9570 | |
Research Use Only | |
GOSR2 | |
Recombinant Protein | |
E. coli | |
MDPLFQQTHK QVHEIQSCMG RLETADKQSV HIVENEIQAS IDQIFSRLER LEILSSKEPP NKRQNARLRV DQLKYDVQHL QTALRNFQHR RHAREQQERQ REELLSRTFT TNDSDTTIPM DESLQFNSSL QKVHNGMDDL ILDGHNILDG LRTQRLTLKG TQKKILDIAN MLGLSNTVMR LIEKRAFQDK | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Golgi SNAP receptor complex member 2 Protein