Få mere at vide
enQuireBio™ Recombinant Human Golgi SNAP receptor complex member 2 Protein
Artikelnummer. 16010850
Mængde:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Pakningsstørrelse:
100µg
10µg
1mg
200µg
500µg
50µg
Artikelnummer. 16010850
Brand: enQuireBio™ QP6107ec1mg
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
A DNA sequence encoding the Homo sapiens (Human) Golgi SNAP receptor complex member 2, was expressed in the hosts and tags indicated.
Tekniske data
Adgangsnummer NP_001012529.1
Gen-id (Entrez) 9570
Navn Golgi SNAP receptor complex member 2 Protein
Mængde 1 mg
Regulatorisk status Research Use Only
Endotoksin koncentration Not determined.
Gen symbol GOSR2
Protein længde Cytoplasmic Domain
Produkttype Recombinant Protein
Krydsreaktivitet Human
Arter E. coli
Protein tag His-SUMO
Sekvens MDPLFQQTHK QVHEIQSCMG RLETADKQSV HIVENEIQAS IDQIFSRLER LEILSSKEPP NKRQNARLRV DQLKYDVQHL QTALRNFQHR RHAREQQERQ REELLSRTFT TNDSDTTIPM DESLQFNSSL QKVHNGMDDL ILDGHNILDG LRTQRLTLKG TQKKILDIAN MLGLSNTVMR LIEKRAFQDK
Buffer Tris-based buffer, 50% glycerol
Renhed eller kvalitetsklasse Greater than 90% as determined by SDS-PAGE.