missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GNAS- subunit alpha isoforms short Protein
A DNA sequence encoding the Homo sapiens (Human) GNAS- subunit alpha isoforms short, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7257-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000507.1 | |
GNAS- subunit alpha isoforms short Protein | |
Research Use Only | |
GNAS | |
Recombinant Protein | |
E. coli | |
MGCLGNSKTE DQRNEEKAQR EANKKIEKQL QKDKQVYRAT HRLLLLGAGE SGKSTIVKQM RILHVNGFNG EGGEEDPQAA RSNSDGEKAT KVQDIKNNLK EAIETIVAAM SNLVPPVELA NPENQFRVDY ILSVMNVPDF DFPPEFYEHA KALWEDEGVR ACYERSNEYQ LIDCAQYFLD KIDVIKQADY VPSDQDLLRC RVLTSGIFET KFQVDKVNFH MFDVGGQRDE RRKWIQCFND VTAIIFVVAS SSYNMVIRED NQTNRLQEAL NLFKSIWNNR WLRTISVILF LNKQDLLAEK VLAGKSKIED YFPEFARYTT PEDATPEPGE DPRVTRAKYF IRDEFLRIST ASGDGRHYCY PHFTCAVDTE NIRRVFNDCR DIIQRMHLRQ YELL | |
Greater than 90% as determined by SDS-PAGE. |
2778 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |