missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GNAS- subunit alpha isoforms short Protein
A DNA sequence encoding the Homo sapiens (Human) GNAS- subunit alpha isoforms short, was expressed in the hosts and tags indicated.
1820.00 DKK - 24645.00 DKK
Specifications
Accession Number | NP_000507.1 |
---|---|
Gene ID (Entrez) | 2778 |
Name | GNAS- subunit alpha isoforms short Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16087890
|
enQuireBio™
QP7257-ec-10ug |
10 ÎĽg | N/A | ||||||
16027900
|
enQuireBio™
QP7257-ec-50ug |
50 ÎĽg | N/A | ||||||
16077890
|
enQuireBio™
QP7257-ec-100ug |
100 ÎĽg |
5555.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16007900
|
enQuireBio™
QP7257-ec-200ug |
200 ÎĽg | N/A | ||||||
16017900
|
enQuireBio™
QP7257-ec-500ug |
500 ÎĽg | N/A | ||||||
16097890
|
enQuireBio™
QP7257-ec-1mg |
1 mg |
24645.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_000507.1 | |
GNAS- subunit alpha isoforms short Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
2778 | |
Research Use Only | |
GNAS | |
Recombinant Protein | |
E. coli | |
MGCLGNSKTE DQRNEEKAQR EANKKIEKQL QKDKQVYRAT HRLLLLGAGE SGKSTIVKQM RILHVNGFNG EGGEEDPQAA RSNSDGEKAT KVQDIKNNLK EAIETIVAAM SNLVPPVELA NPENQFRVDY ILSVMNVPDF DFPPEFYEHA KALWEDEGVR ACYERSNEYQ LIDCAQYFLD KIDVIKQADY VPSDQDLLRC RVLTSGIFET KFQVDKVNFH MFDVGGQRDE RRKWIQCFND VTAIIFVVAS SSYNMVIRED NQTNRLQEAL NLFKSIWNNR WLRTISVILF LNKQDLLAEK VLAGKSKIED YFPEFARYTT PEDATPEPGE DPRVTRAKYF IRDEFLRIST ASGDGRHYCY PHFTCAVDTE NIRRVFNDCR DIIQRMHLRQ YELL | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human GNAS- subunit alpha isoforms short Protein