missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Hygromycin-B 4-O-kinase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Hygromycin-B 4-O-kinase, was expressed in the hosts and tags indicated.
2615.00 DKK - 23570.00 DKK
Specifications
Accession Number | WP_000742814.1 |
---|---|
Gene ID (Entrez) | 13905269 |
Name | Hygromycin-B 4-O-kinase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16098660
|
enQuireBio™
QP7397-ye-10ug |
10 ÎĽg | N/A | ||||||
16038670
|
enQuireBio™
QP7397-ye-50ug |
50 ÎĽg | N/A | ||||||
16088660
|
enQuireBio™
QP7397-ye-100ug |
100 ÎĽg |
7930.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16018670
|
enQuireBio™
QP7397-ye-200ug |
200 ÎĽg | N/A | ||||||
16028670
|
enQuireBio™
QP7397-ye-500ug |
500 ÎĽg |
19165.00 DKK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16008670
|
enQuireBio™
QP7397-ye-1mg |
1 mg | N/A | ||||||
Specifications
WP_000742814.1 | |
Hygromycin-B 4-O-kinase Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
13905269 | |
Research Use Only | |
hph | |
Recombinant Protein | |
Yeast | |
MKKPELTATS VEKFLIEKFD SVSDLMQLSE GEESRAFSFD VGGRGYVLRV NSCADGFYKD RYVYRHFASA ALPIPEVLDI GEFSESLTYC ISRRAQGVTL QDLPETELPA VLQPVAEAMD AIAAADLSQT SGFGPFGPQG IGQYTTWRDF ICAIADPHVY HWQTVMDDTV SASVAQALDE LMLWAEDCPE VRHLVHADFG SNNVLTDNGR ITAVIDWSEA MFGDSQYEVA NIFFWRPWLA CMEQQTRYFE RRHPELAGSP RLRAYMLRIG LDQLYQSLVD GNFDDAAWAQ GRCDAIVRSG AGTVGRTQIA RRSAAVWTDG CVEVLADSGN RRPSTRPRAK E | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant E. coli Hygromycin-B 4-O-kinase Protein