missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Glc operon transcriptional activator Protein
A DNA sequence encoding the Escherichia coli O6:H1 (strain CFT073 / ATCC 700928 / UPEC) Glc operon transcriptional activator, was expressed in the hosts and tags indicated.
2300.00 DKK - 24645.00 DKK
Specifications
Accession Number | WP_001297764.1 |
---|---|
Name | Glc operon transcriptional activator Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | glcC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16078650
|
enQuireBio™
QP7395-ec-10ug |
10 ÎĽg | N/A | ||||||
16018660
|
enQuireBio™
QP7395-ec-50ug |
50 ÎĽg | N/A | ||||||
16068650
|
enQuireBio™
QP7395-ec-100ug |
100 ÎĽg |
7110.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16098650
|
enQuireBio™
QP7395-ec-200ug |
200 ÎĽg | N/A | ||||||
16008660
|
enQuireBio™
QP7395-ec-500ug |
500 ÎĽg | N/A | ||||||
16088650
|
enQuireBio™
QP7395-ec-1mg |
1 mg |
24645.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
WP_001297764.1 | |
Research Use Only | |
glcC | |
Recombinant Protein | |
E. coli | |
MKDERRPICE VVAESIERLI IDGVLKVGQP LPSERRLCEK LGFSRSALRE GLTVLRGRGI IETAQGRDSR VARLNRVQDT SPLIHLFSTQ PRTLYDLLDV RALLEGESAR LAATLGTQAD FVVITRCYEK MLAASENNKE ISLIEHAQLD HAFHLAICQA SHNQVLVFTL QSLTDLMFNS VFASVNNLYH RPQQKKQIDR QHARIYNAVL QRLPHVAQRA ARDHVRTVKK NLHDIELEGH HLIRSAVPLE MNLS | |
Greater than 90% as determined by SDS-PAGE. |
Glc operon transcriptional activator Protein | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant E. coli Glc operon transcriptional activator Protein