missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Glc operon transcriptional activator Protein
A DNA sequence encoding the Escherichia coli O6:H1 (strain CFT073 / ATCC 700928 / UPEC) Glc operon transcriptional activator, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7395-ec-10ug
This item is not returnable.
View return policy
Specifications
WP_001297764.1 | |
10 ÎĽg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Glc operon transcriptional activator Protein | |
Research Use Only | |
glcC | |
Recombinant Protein | |
E. coli | |
MKDERRPICE VVAESIERLI IDGVLKVGQP LPSERRLCEK LGFSRSALRE GLTVLRGRGI IETAQGRDSR VARLNRVQDT SPLIHLFSTQ PRTLYDLLDV RALLEGESAR LAATLGTQAD FVVITRCYEK MLAASENNKE ISLIEHAQLD HAFHLAICQA SHNQVLVFTL QSLTDLMFNS VFASVNNLYH RPQQKKQIDR QHARIYNAVL QRLPHVAQRA ARDHVRTVKK NLHDIELEGH HLIRSAVPLE MNLS | |
Greater than 90% as determined by SDS-PAGE. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant E. coli Glc operon transcriptional activator Protein > 10μg
Spot an opportunity for improvement?Share a Content Correction