missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli ATP-dependent DNA helicase Rep Protein

A DNA sequence encoding the Escherichia coli (strain K12) ATP-dependent DNA helicase Rep, was expressed in the hosts and tags indicated.

2300.00 DKK - 24645.00 DKK

Specifications

Accession Number WP_001238899.1
Gene ID (Entrez) 948292
Name ATP-dependent DNA helicase Rep Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16075851
View Documents
enQuireBio™
QP8666-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16015861
View Documents
enQuireBio™
QP8666-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16065851
View Documents
enQuireBio™
QP8666-ec-100ug
100 ÎĽg
7110.00 DKK
100µg
Please to purchase this item. Need a web account? Register with us today!
16095851
View Documents
enQuireBio™
QP8666-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16005861
View Documents
enQuireBio™
QP8666-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16085851
View Documents
enQuireBio™
QP8666-ec-1mg
1 mg
24645.00 DKK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant E. coli ATP-dependent DNA helicase Rep Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.