missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 50S ribosomal protein L28 Protein
A DNA sequence encoding the Escherichia coli (strain K12) 50S ribosomal protein L28, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8636-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_418094.1 | |
50S ribosomal protein L28 Protein | |
Research Use Only | |
rpmB | |
Recombinant Protein | |
E. coli | |
SRVCQVTGKR PVTGNNRSHA LNATKRRFLP NLHSHRFWVE SEKRFVTLRV SAKGMRVIDK KGIDTVLAEL RARGEKY | |
Greater than 90% as determined by SDS-PAGE. |
946941 | |
500 μg | |
Not determined. | |
Full Length | |
E. coli | |
GST | |
Tris-based buffer, 50% glycerol |