missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 30S ribosomal protein S18 Protein
A DNA sequence encoding the Escherichia coli (strain K12) 30S ribosomal protein S18, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8618-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_418623.1 | |
30S ribosomal protein S18 Protein | |
Research Use Only | |
rpsR | |
Recombinant Protein | |
E. coli | |
ARYFRRRKFC RFTAEGVQEI DYKDIATLKN YITESGKIVP SRITGTRAKY QRQLARAIKR ARYLSLLPYT DRHQ | |
Greater than 90% as determined by SDS-PAGE. |
948721 | |
10 μg | |
Not determined. | |
Full Length | |
E. coli | |
GST | |
Tris-based buffer, 50% glycerol |