missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Cytomegalovirus Uncharacterized protein UL131 Protein
A DNA sequence encoding the Human cytomegalovirus (strain AD169) (HHV-5) (Human herpesvirus 5) Uncharacterized protein UL131, was expressed in the hosts and tags indicated.
3490.00 DKK - 27120.00 DKK
Specifications
Name | Uncharacterized protein UL131 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | UL131 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16087340
|
enQuireBio™
QP7086-iv-10ug |
10 ÎĽg | N/A | ||||||
16097340
|
enQuireBio™
QP7086-iv-50ug |
50 ÎĽg | N/A | ||||||
16077340
|
enQuireBio™
QP7086-iv-100ug |
100 ÎĽg |
12320.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16027340
|
enQuireBio™
QP7086-200ug |
200 ÎĽg |
12775.00 DKK
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16037340
|
enQuireBio™
QP7086-500ug |
500 ÎĽg |
17065.00 DKK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16017340
|
enQuireBio™
QP7086-1mg |
1 mg |
27120.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Uncharacterized protein UL131 Protein | |
Not determined. | |
Full Length | |
Cytomegalovirus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
UL131 | |
Recombinant Protein | |
E. coli | |
MCMMSHNKAF FLSLQHAAVS GVAVCLSVRR GAGSVPAGNR GKKTIITEYR ITGTRALARC PTKPVTSMWN SSWTSR | |
Greater than 80% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Cytomegalovirus Uncharacterized protein UL131 Protein