missing translation for 'onlineSavingsMsg'
Få mere at vide

enQuireBio™ Recombinant Cytomegalovirus Uncharacterized protein UL131 Protein

Artikelnummer. 16097340
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
missing translation for 'unitSize'
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16097340

missing translation for 'mfr': enQuireBio™ QP7086iv50ug

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy

A DNA sequence encoding the Human cytomegalovirus (strain AD169) (HHV-5) (Human herpesvirus 5) Uncharacterized protein UL131, was expressed in the hosts and tags indicated.

Specifications

Navn Uncharacterized protein UL131 Protein
Mængde 50 μg
Regulatorisk status Research Use Only
Endotoksin koncentration Not determined.
Gen symbol UL131
Protein længde Full Length
Produkttype Recombinant Protein
Krydsreaktivitet Cytomegalovirus
Arter E. coli
Protein tag His-SUMO
Sekvens MCMMSHNKAF FLSLQHAAVS GVAVCLSVRR GAGSVPAGNR GKKTIITEYR ITGTRALARC PTKPVTSMWN SSWTSR
Buffer Tris-based buffer, 50% glycerol
Renhed eller kvalitetsklasse Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.