missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant A. pasturia Modification methylase ApaLI Protein

A DNA sequence encoding the Acetobacter pasteurianus (Acetobacter turbidans) Modification methylase ApaLI, was expressed in the hosts and tags indicated.

2300.00 DKK - 24645.00 DKK

Specifications

Name Modification methylase ApaLI Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol apaLIM
Protein Length Full Length
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16078950
View Documents
enQuireBio™
QP7481-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16018960
View Documents
enQuireBio™
QP7481-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16068950
View Documents
enQuireBio™
QP7481-ec-100ug
100 ÎĽg
7110.00 DKK
100µg
Please to purchase this item. Need a web account? Register with us today!
16098950
View Documents
enQuireBio™
QP7481-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16008960
View Documents
enQuireBio™
QP7481-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16088950
View Documents
enQuireBio™
QP7481-ec-1mg
1 mg
24645.00 DKK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Modification methylase ApaLI Protein
Not determined.
Full Length
A. pasturia
His-SUMO
Tris-based buffer, 50% glycerol
Research Use Only
apaLIM
Recombinant Protein
E. coli
MNKDEVVVSL FAGAGGFSSG FSQAGLKPLF GAEINADACQ TYQENVGSPC HQLDLSTVDP SHIEMLTGGK RPFVVIGGPP CQGFSTAGPR NFADPRNLLI FNYLNIVERL SPRWLIFENV EGLLTSGGGR DLARLVREFV DMGYSVRLQK VNLAAYGVPQ TRKRVLIIGN RLGIDFQFPE ELYSFDSGKA KKASGKPLAP SLAEAVAGLG PAASDKDALV PYASSEPVNA FDARMRAGNR VEVVTHHVRV EAAERMQVEL LKPGQTMKDL PPELWHESYR RRANRRVSDG TPTEKRGGAP SGIKRLHGNL QSLTITGPAA REFIHPTEHR PLTIRECARI QTFPDKYRWV GNNASVIQQI GNAVPPLAAE RLAKHLRDID GSFGADTRPA GAMSAKLLGF VLTEALGMSP ALKSTEALLA EMHQGGFVF
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant A. pasturia Modification methylase ApaLI Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.