Få mere at vide
enQuireBio™ Recombinant A. pasturia Modification methylase ApaLI Protein
Artikelnummer. 16008960
Mængde:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Pakningsstørrelse:
100µg
10µg
1mg
200µg
500µg
50µg
Artikelnummer. 16008960
Brand: enQuireBio™ QP7481ec500ug
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
A DNA sequence encoding the Acetobacter pasteurianus (Acetobacter turbidans) Modification methylase ApaLI, was expressed in the hosts and tags indicated.
Tekniske data
Navn Modification methylase ApaLI Protein
Mængde 500 μg
Regulatorisk status Research Use Only
Endotoksin koncentration Not determined.
Gen symbol apaLIM
Protein længde Full Length
Produkttype Recombinant Protein
Krydsreaktivitet A. pasturia
Arter E. coli
Protein tag His-SUMO
Sekvens MNKDEVVVSL FAGAGGFSSG FSQAGLKPLF GAEINADACQ TYQENVGSPC HQLDLSTVDP SHIEMLTGGK RPFVVIGGPP CQGFSTAGPR NFADPRNLLI FNYLNIVERL SPRWLIFENV EGLLTSGGGR DLARLVREFV DMGYSVRLQK VNLAAYGVPQ TRKRVLIIGN RLGIDFQFPE ELYSFDSGKA KKASGKPLAP SLAEAVAGLG PAASDKDALV PYASSEPVNA FDARMRAGNR VEVVTHHVRV EAAERMQVEL LKPGQTMKDL PPELWHESYR RRANRRVSDG TPTEKRGGAP SGIKRLHGNL QSLTITGPAA REFIHPTEHR PLTIRECARI QTFPDKYRWV GNNASVIQQI GNAVPPLAAE RLAKHLRDID GSFGADTRPA GAMSAKLLGF VLTEALGMSP ALKSTEALLA EMHQGGFVF
Buffer Tris-based buffer, 50% glycerol
Renhed eller kvalitetsklasse Greater than 90% as determined by SDS-PAGE.