missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant A. baumannii Elongation factor Tu Protein
A DNA sequence encoding the Acinetobacter baumannii (strain ATCC 17978 / CIP 53.77 / LMG 1025 / NCDC KC755 / 5377) Elongation factor Tu, was expressed in the hosts and tags indicated.
2615.00 DKK - 23570.00 DKK
Specifications
Name | Elongation factor Tu Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | tuf1 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16056310
|
enQuireBio™
QP6854-ye-10ug |
10 ÎĽg | N/A | ||||||
16096310
|
enQuireBio™
QP6854-ye-50ug |
50 ÎĽg | N/A | ||||||
16046310
|
enQuireBio™
QP6854-ye-100ug |
100 ÎĽg |
7930.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16076310
|
enQuireBio™
QP6854-ye-200ug |
200 ÎĽg | N/A | ||||||
16086310
|
enQuireBio™
QP6854-ye-500ug |
500 ÎĽg |
19165.00 DKK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16066310
|
enQuireBio™
QP6854-ye-1mg |
1 mg | N/A | ||||||
Specifications
Elongation factor Tu Protein | |
Not determined. | |
Full Length | |
A. baumannii | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
tuf1 | |
Recombinant Protein | |
Yeast | |
MAKAKFERNK PHVNVGTIGH VDHGKTTLTA AIATICAKTY GGEAKDYSQI DSAPEEKARG ITINTSHVEY DSPTRHYAHV DCPGHADYVK NMITGAAQMD GAILVCAATD GPMPQTREHI LLSRQVGVPY IIVFLNKCDL VDDEELLELV EMEVRELLST YDFPGDDTPV IRGSALAALN GEAGPYGEES VLALVAALDS YIPEPERAID KAFLMPIEDV FSISGRGTVV TGRVEAGIIK VGEEVEIVGI KDTVKTTVTG VEMFRKLLDE GRAGENCGIL LRGTKREEVQ RGQVLAKPGT IKPHTKFDAE VYVLSKEEGG RHTPFLNGYR PQFYFRTTDV TGAIQLKEGV EMVMPGDNVE MSVELIHPIA MDPGLRFAIR EGGRTVGAGV VAKVTA | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant A. baumannii Elongation factor Tu Protein