missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant A. baumannii Elongation factor Tu Protein
A DNA sequence encoding the Acinetobacter baumannii (strain ATCC 17978 / CIP 53.77 / LMG 1025 / NCDC KC755 / 5377) Elongation factor Tu, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6854-ye-50ug
This item is not returnable.
View return policy
Specifications
Elongation factor Tu Protein | |
Research Use Only | |
tuf1 | |
Recombinant Protein | |
Yeast | |
MAKAKFERNK PHVNVGTIGH VDHGKTTLTA AIATICAKTY GGEAKDYSQI DSAPEEKARG ITINTSHVEY DSPTRHYAHV DCPGHADYVK NMITGAAQMD GAILVCAATD GPMPQTREHI LLSRQVGVPY IIVFLNKCDL VDDEELLELV EMEVRELLST YDFPGDDTPV IRGSALAALN GEAGPYGEES VLALVAALDS YIPEPERAID KAFLMPIEDV FSISGRGTVV TGRVEAGIIK VGEEVEIVGI KDTVKTTVTG VEMFRKLLDE GRAGENCGIL LRGTKREEVQ RGQVLAKPGT IKPHTKFDAE VYVLSKEEGG RHTPFLNGYR PQFYFRTTDV TGAIQLKEGV EMVMPGDNVE MSVELIHPIA MDPGLRFAIR EGGRTVGAGV VAKVTA | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Full Length | |
A. baumannii | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant A. baumannii Elongation factor Tu Protein > 50μg
Spot an opportunity for improvement?Share a Content Correction