Learn More
Heidolph™ Receiving Flask
For use with rotary evaporators
Brand: Heidolph™ 15-300-004-11
This item is not returnable.
View return policy
Description
- With S 35/20
- Part number 15-300-004-11: includes drain valves
- Included as a standard for all Hei-VAP Industrial models
Specifications
Clear | |
Receiving Flask | |
35/20 |
Glass | |
Rotary Evaporators | |
10 L |
Product Content Correction
Product Title
Heidolph™ Receiving Flask > Receiving flasks 10 L; with drain valves
Spot an opportunity for improvement?Share a Content Correction