Learn More
Heidolph Instruments Receiving flask
For use with rotary evaporators
Brand:  Heidolph Instruments 514-85000-00
 View more versions of this product
Product Code. 10259601
  • 923.00 DKK / Each
Estimated Shipment: 11-06-2025
Description
  • With S 35/20
  • Part number 15-300-004-11: includes drain valves
    • Included as a standard for all Hei-VAP Industrial models
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Non-coated
Evaporating Flask
2000 mL
Borosilicate Glass
S 35/20
115/230 VAC
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Heidolph Instruments Receiving flask > Capacity: 2000 mL