missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Rat CXCL1/GRO alpha/KC/CINC-1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
1345.00 DKK - 3110.00 DKK
Tekniske data
| Antigen | CXCL1/GRO alpha/KC/CINC-1 |
|---|---|
| Klon | 2176C |
| Fortynding | Western Blot 2 ug/mL, Neutralization 0.075-0.45 ug/mL |
| Applikationer | Western Blot, Neutralization |
| Klassifikation | Monoclonal |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
18754228
![]() |
R&D Systems
MAB5151-SP |
25 μg |
1345.00 DKK
25µg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
18704768
![]() |
R&D Systems
MAB5151-100 |
100 μg |
3110.00 DKK
100µg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
CXCL1/GRO alpha/KC/CINC-1 Monoclonal specifically detects CXCL1/GRO alpha/KC/CINC-1 in Rat samples. It is validated for Western Blot, Neutralization.Tekniske data
| CXCL1/GRO alpha/KC/CINC-1 | |
| Western Blot 2 ug/mL, Neutralization 0.075-0.45 ug/mL | |
| Monoclonal | |
| Supernatant | |
| Rat | |
| P14095 | |
| 2919 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 2176C | |
| Western Blot, Neutralization | |
| Unconjugated | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1 | |
| E. coli-derived recombinant rat CXCL1/GRO alpha/KC/CINC-1 Ala25-Lys96 Accession # P14095 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects rat CXCL1/GRO alpha/KC/CINC-1 in direct ELISAs. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel