missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Rat CXCL1/GRO alpha/KC/CINC-1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB5151-100
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CXCL1/GRO alpha/KC/CINC-1 Monoclonal specifically detects CXCL1/GRO alpha/KC/CINC-1 in Rat samples. It is validated for Western Blot, Neutralization.
Tekniske data
| CXCL1/GRO alpha/KC/CINC-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1 | |
| E. coli-derived recombinant rat CXCL1/GRO alpha/KC/CINC-1 Ala25-Lys96 Accession # P14095 | |
| 100 μg | |
| 2919 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Neutralization | |
| 2176C | |
| Western Blot 2 ug/mL, Neutralization 0.075-0.45 ug/mL | |
| P14095 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects rat CXCL1/GRO alpha/KC/CINC-1 in direct ELISAs. | |
| Rat | |
| Supernatant |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion