Få mere at vide
Raltegravir Beta-D-Glucuronide, TRC
Artikelnummer. 30005117
Skift visning
Mængde:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30005117 1 mg
30005444 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30005117 Leverandør Toronto Research Chemicals Leverandørnr. TRCR1003101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 952654-62-5
Molekylær formel C26 H29 F N6 O11
Molekylvægt (g/mol) 620.54
Synonym Raltegravir-β-D-Glucuronide
IUPAC navn (2S,3S,4S,5R,6S)-6-[4-[(4-fluorophenyl)methylcarbamoyl]-1-methyl-2-[2-[(5-methyl-1,3,4-oxadiazole-2-carbonyl)amino]propan-2-yl]-6-oxopyrimidin-5-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
SMIL CN1C(=O)C(=C(N=C1C(C)(C)NC(=O)c2oc(C)nn2)C(=O)NCc3ccc(F)cc3)O[C@@H]4O[C@@H]([C@@H](O)[C@H](O)[C@H]4O)C(=O)O
Tekniske data
Kemisk navn eller materiale Raltegravir beta-D-glucuronide
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C26H29FN6O11/c1-10-31-32-21(42-10)20(38)30-26(2,3)25-29-13(19(37)28-9-11-5-7-12(27)8-6-11)17(22(39)33(25)4)43-24-16(36)14(34)15(35)18(44-24)23(40)41/h5-8,14-16,18,24,34-36H,9H2,1-4H3,(H,28,37)(H,30,38)(H,40,41)/t14-,15-,16+,18-,24+/m0/s1
Formel vægt 620.1878
Procent renhed >95%