missing translation for 'onlineSavingsMsg'
Få mere at vide

Rafoxanide, TRC

Artikelnummer. 17963305
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17963305 250 mg
17911615 500 mg
17932265 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17963305 Leverandør Toronto Research Chemicals Leverandørnr. TRCR073500250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22662-39-1
Molekylær formel C19 H11 Cl2 I2 N O3
Molekylvægt (g/mol) 626.01
Synonym Benzamide, N-[3-chloro-4-(4-chlorophenoxy)phenyl]-2-hydroxy-3,5-diiodo-, Salicylanilide, 3'-chloro-4'-(p-chlorophenoxy)-3,5-diiodo- (8CI), 3,5-Diiodo-3'-chloro-4'-(p-chlorophenoxy)salicylanilide, 3,5-Diodo-3'-chloro-4'-(p-chlorophenoxy)salicylanilide, 3'-Chloro-4'-(4-chlorophenoxy)-3,5-diiodosalicylanilide, 3'-Chloro-4'-(p-chlorophenoxy)-3,5-diiodosalicylanilide, Disalan, NSC 355278, Rafoxanide, Ranide
IUPAC navn N-[3-chloro-4-(4-chlorophenoxy)phenyl]-2-hydroxy-3,5-diiodobenzamide
SMIL Oc1c(I)cc(I)cc1C(=O)Nc2ccc(Oc3ccc(Cl)cc3)c(Cl)c2

Specifications

Kemisk navn eller materiale Rafoxanide
Renhedsgrad noter HPLC
Mængde 250 mg
InChI formel InChI=1S/C19H11Cl2I2NO3/c20-10-1-4-13(5-2-10)27-17-6-3-12(9-15(17)21)24-19(26)14-7-11(22)8-16(23)18(14)25/h1-9,25H,(H,24,26)
Formel vægt 624.8205
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.