Learn More
Spectrum Labs™ Rack Set for CF-2 Fraction Collector
Spectrum Labs™ Rack Set for CF-2 Fraction Collector is a full set of racks for one specifically sized tube designed for use with the versatile CF-2 collector.
6205.00 DKK - 6465.00 DKK
Specifications
Type | Fraction Collector Tube Rack |
---|---|
For Use With | CF-2 Fraction Collector |
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
11959578
|
Spectrum Labs™
124855 |
Holds 42 x 28 mm diameter scintilation vials | |||||||
11969578
|
Spectrum Labs™
124856 |
Holds 116 x 17 mm or 18 mm diameter tubes | |||||||
Description
- Designed for use with the versatile CF-2 Fraction Collector
- Complete set of tube racks designed to accommodate the tubes chosen
Specifications
Fraction Collector Tube Rack | |
CF-2 Fraction Collector |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Spectrum Labs™ Rack Set for CF-2 Fraction Collector