Få mere at vide
rac Terpinen-4-ol, TRC
Artikelnummer. 30009524
Skift visning
Mængde:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009524 500 mg
30007902 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30009524 Leverandør Toronto Research Chemicals Leverandørnr. TRCT117500500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 562-74-3
Molekylær formel C10 H18 O
Molekylvægt (g/mol) 154.25
Synonym 3-Cyclohexen-1-ol, 4-methyl-1-(1-methylethyl)-, p-Menth-1-en-4-ol (8CI), 4-Methyl-1-(1-methylethyl)-3-cyclohexen-1-ol, (±)-4-Terpineol, (±)-Terpinen-4-ol, 1-(1-Methylethyl)-4-methyl-3-cyclohexen-1-ol, 1-Isopropyl-4-methylcyclohex-3-en-1-ol, 1-Terpen-4-ol, 1-Terpinen-4-ol, 4-Carvomenthenol, 4-Methyl-1-(1-methylethyl)-3-cyclohexene-1-ol, 4-Methyl-1-isopropyl-3-cyclohexen-1-ol, 4-Terpinenol, 4-Terpineol, Melaleucol, NSC 147749, Terpin-4-ol, Terpinen-4-ol, Terpinene-4-ol, Terpineol-4, dl-4-Terpineol
IUPAC navn 4-methyl-1-propan-2-ylcyclohex-3-en-1-ol
SMIL CC(C)C1(O)CCC(=CC1)C
Tekniske data
Kemisk navn eller materiale 1-Terpinen-4-ol
Mængde 500 mg
InChI formel InChI=1S/C10H18O/c1-8(2)10(11)6-4-9(3)5-7-10/h4,8,11H,5-7H2,1-3H3
Formel vægt 154.1358